Learn More
Quickfit™ Expansion Socket Adapter
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Brand: Quickfit XA31
36.51 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
- Useful for the connection of dissimilar sized ground glass joints
- Large sized socket at the top and small sized cone at the base
Specifications
14/23 | |
Borosilicate Glass | |
24/29 |
Useful for the connection of dissimilar sized ground Glass joints | |
Expansion adapter |
Product information
File size: 2.8 MB
Product Content Correction
Product Title
Quickfit™ Expansion Socket Adapter > Cone size: 14/23; Socket size: 24/29
Spot an opportunity for improvement?Share a Content Correction