missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Female Spherical Ball Joint
For use with male ball joints MS:
Brand: Quickfit FS13UB
204.25 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with male ball joints MS:
- Female cup joints precision manufactured to the highest standards
- Spherical joints are manufactured with an approximate shank length of 100mm
Specifications
Female Spherical Ball Joint | |
S13 | |
0.05in. | |
1.5mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction