missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Flat Flange Joint

Ideal for use as pipeline joints or in the construction of flat flange lids for use with wide neck reaction flasks.

48.00 € - 510.00 €


Products 4
Product Code Brand Thickness (Metric) Wall Price Quantity & Availability  
Product Code Brand Thickness (Metric) Wall Price Quantity & Availability  
12448956
View Documents
Quickfit
FG25
1.8mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12091712
View Documents
Quickfit
FG100
4mm
97.85 €
Each
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
 
15577840
View Documents
Quickfit
FG50
2.2mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15587840
View Documents
Quickfit
FG75
4.4mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Highest quality flat flange joints
  • Precision ground for leak free performance
  • Choice of sizes up to 100mm bore
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.