Learn More
Quickfit™ Glass Conical Funnel with Glass Sintered Disc
Constructed of chemically resistant borosilicate glass. Quickfit™ Glass Conical Funnel with Glass Sintered Disc is resistant to most corrosive reagents such as ammonia and sulphuric acid. Conical shape with glass sintered disc eliminates the need for filter papers. Integral ground glass cone on the stem connects to Quickfit™ Büchner flasks and vessels.
Brand: Quickfit SF4B32
95.95 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Constructed of chemically resistant borosilicate glass. Quickfit™ Glass Conical Funnel with Glass Sintered Disc is resistant to most corrosive reagents such as ammonia and sulphuric acid. Conical shape with glass sintered disc eliminates the need for filter papers. Integral ground glass cone on the stem connects to Quickfit™ Büchner flasks and vessels.
- Conical shape with glass sintered disc
- Resistant to most corrosive reagents - Ammonia, sulfuric acid, etc
- With integral ground glass cone on the stem for connecting to Quickfit™ Büchner flasks
Specifications
Borosilicate Glass | |
For connecting to Quickfit™ Buchner flasks | |
Sintered glass disc | |
16 to 40μm |
19/26 | |
30mm | |
3 | |
Filter Funnel |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.