missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Glass Funnel with Rotaflo™ Stopcock and Open Top

Enables easy fitting to the top of Quickfit™ chromatography columns etc where protruding stem would be undesirable.

Brand:  Quickfit D3/12

Product Code. 15567790

  • 662.00 € / Pack of 5
Estimated Shipment: 01-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Complies with ISO 4800, Type 3
  • With stem inside the cone
  • GP Rotaflo™ stopcock with PTFE key for grease-free performance
  • With open top instead of socket for easy addition of solutions
TRUSTED_SUSTAINABILITY
Specifications

Specifications

28mm
Borosilicate Glass
19/26
Dropping Funnel
50 mL
ISO 4800
GP Rotaflo™ stopcock, PTFE key
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Quickfit™ Glass Funnel with Rotaflo™ Stopcock and Open Top > Capacity: 50mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.