missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Heating Element

Quickfit™ Heating Element is a spare part for use with QSW4 Series Water Still.

Brand:  Scilabware QWSH4/220V

498.75 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15898756

  • 554.00 € / 1 unit
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Chromium plated heating element
  • Includes 2 independent re-settable thermostats
  • Protect the still in the event of water supply failure
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50/60 Hz
Chromium-plated
3 kW
Heating Element
2 Thermostats
220 V
QWS4 Series Water Still
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers