Learn More
Quickfit™ Hexagonal Hollow-blown Glass Stopper
Ideally suited for use with volumetric flasks.
74.70 € - 125.00 €
Specifications
For Use With (Equipment) Volumetric flasks
View More Specs
Products 9
Product Code Brand Standard Taper Joint Price Quantity & Availability  
Product Code Brand Standard Taper Joint Price Quantity & Availability  
12421788
View Documents
Quickfit
TRF691
10/19
96.45 €
Pack of 10
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
13160063
View Documents
Quickfit
TRF693
14/23
115.00 €
Pack of 10
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
12491788
View Documents
Quickfit
TRF699
45/40 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12481788
View Documents
Quickfit
TRF698
NS 34/35
102.00 €
Pack of 5
Estimated Shipment: 10-04-2025
to see stock.
Add to basket
12471788
View Documents
Quickfit
TRF697
NS 29/32
120.00 €
Pack of 10
Estimated Shipment: 10-04-2025
to see stock.
Add to basket
12461788
View Documents
Quickfit
TRF696
NS 24/29
104.00 €
Pack of 10
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
11339093
View Documents
Quickfit
TRF690
NS 7/16
74.70 €
Pack of 10
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
12451788
View Documents
Quickfit
TRF695
NS 19/26
125.00 €
Pack of 10
Estimated Shipment: 10-04-2025
to see stock.
Add to basket
13150063
View Documents
Quickfit
TRF692
NS 12/21
96.10 €
Pack of 10
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
Description
Description
Ideally suited for use with volumetric flasks.
  • Lower closed end of stopper features 'drip-tip' to ensure adhered liquid is returned to flask
  • Hexagonal head prevents stopper from rolling and provides good grip
  • Flat head allows stopper to be stood on end, thus preventing contamination of ground surface
  • Hollow-blown to reduce weight
Specifications
Specifications
Volumetric flasks