Learn More
Pyrex™ Quickfit™ Metal Retaining Clips
Use with soxhlet extractors, flat flange lids and reaction vessels. Pyrex™ Quickfit™ Metal Retaining Clips apply positive pressure to both surfaces with spring wire design.
47.05 € - 56.80 €
Specifications
Material Stainless Steel
For Use With (Application) Securing flat flange lids to wide neck reaction flasks and culture vessels
Description Spring wire design
View More Specs
Products 3
Product Code Brand Size Diameter (Metric) Inside Price Quantity & Availability  
Product Code Brand Size Diameter (Metric) Inside Price Quantity & Availability  
11312015
View Documents
Pyrex™
JC100F
FG100 100 mm
56.80 €
Each
In Stock
to see stock.
Edge
Add to basket
12405286
View Documents
Pyrex™
JC50F
FG50 50 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10166304
View Documents
Pyrex™
JC75F
FG75 75 mm
47.20 €
Each
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
Description
Description
Use with soxhlet extractors, flat flange lids and reaction vessels. Pyrex™ Quickfit™ Metal Retaining Clips apply positive pressure to both surfaces with spring wire design.
  • For securing flat flange lids to wide neck reaction flasks and culture vessels
  • Stainless steel construction
Specifications
Specifications
Stainless Steel
Securing flat flange lids to wide neck reaction flasks and culture vessels
Spring wire design