missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Pear-Shaped Glass Funnel with Glass Key and Stem with Cone

For use in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.

Brand:  Quickfit D2/11

116.85 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12000083

  • 127.00 € / Each
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.

  • Complies with ISO 4800 Type 2
  • Funnel stem with cone to allow easy connection to Quickfit™ flasks etc
  • Stopcock features interchangeable glass key
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50mm
Borosilicate Glass
14/23
1PK
Separatory Funnel
50 mL
ISO 4800
14/23
1.5mm; Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.