missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Plain Digestion Tubes
For use with block heaters
Brand: Quickfit TT100/26
137.75 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- With plain beaded neck
- Manufactured from borosilicate glass for highest chemical and thermal durability
Specifications
Digestion Tube | |
Borosilicate Glass (3.3) | |
300 mm |
Capacity: 100mL | |
26 mm |