Learn More
Quickfit™ Reaction Vessel Bubbler
Ideal for closing reaction mixture from the atmosphere.
Brand:  Quickfit GB1/19
 View more versions of this product
Product Code. 15537910
  • 887.00 € / Pack of 5
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Side arm to facilitate filling with mineral oil
  • Bubblers have capacity of approx 5ml
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reaction Vessel Bubbler
Ideal for closing reaction mixture from the atmosphere