Learn More
Quickfit™ Reaction Vessel Bubbler
Ideal for closing reaction mixture from the atmosphere.
Brand:  Quickfit GB1/29
 View more versions of this product
Product Code. 15778017
  • 194.00 € / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Side arm to facilitate filling with mineral oil
  • Bubblers have capacity of approx 5ml
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reaction Vessel Bubbler
29/32