missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Silicone Rings for Screw Thread Glass Joints

For use with Quickfit™ glass screw threads

Brand:  Quickfit QR28/6

87.88 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11302085

  • 94.85 € / Pack of 10
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use with Quickfit™ glass screwthreads

  • For use with Quickfit™ glass screwthreads
  • Melamine screwcaps with aperture at the top
  • Silicone rubber rings with PTFE washers
To determine what components are required: i) Check the size of the glass screwthread (size designation refers to O.D. of thread in mm) ii) Check the O.D. of the sampling tube, thermometer, probe etc. that is to be passed through the screwcap iii) Remember to order relevant cap, washer and ring separately
TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 to 7mm
Quickfit™ glass screw threads
Thread size: 28mm
Silicone Rubber Ring
For Use with Quickfit Glass Screw threads
10 Pack
Silicone
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers