missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Silicone Rings for Screw Thread Glass Joints
For use with Quickfit™ glass screw threads
Brand: Quickfit QR24/6
51.97 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with Quickfit™ glass screwthreads
- For use with Quickfit™ glass screwthreads
- Melamine screwcaps with aperture at the top
- Silicone rubber rings with PTFE washers
Specifications
6 to 7mm | |
Thread size: 24mm | |
Silicone Rubber Ring |
Quickfit™ glass screw threads | |
Silicone |