Learn More
Quickfit™ Sloping Splash Head Adapter
Click to view available options
Cone Size:
19/26 (Condenser), 19/26 (Flask)
19/26 (Condenser), 24/29 (Flask)
24/29 (Condenser), 24/29 (Flask)
29/32 (Condenser), 29/32 (Flask)
Unit Size:
Pack of 5
Description
- Used in distillation assemblies to prevent raw liquid carrying over from flask to condenser
Specifications
Specifications
| Material | Borosilicate Glass |
| For Use With (Application) | Distillation assemblies to prevent raw liquid 'carrying over' from flask to condenser |
| Cone Size | 19/26 (Condenser), 19/26 (Flask) |
| Product Type | Sloping Splash Head Adapter |