Learn More
SVL™ Socket/Screw Joint Adapter
Facilitates connection between SVL™ and Quickfit™ glassware.
313.00 € - 826.00 €
Specifications
For Use With (Equipment) | SVL™ and Quickfit™ Glassware |
---|---|
Material | Borosilicate Glass |
Product Type | Socket/screw thread ground joint adapter |
Description
- Features an SVL™ screwthread and Quickfit™ ground glass socket
- Thread accepts SVL™ screw caps and couplings. Socket fits Quickfit™ cones
- Manufactured from temperature and chemical resistant borosilicate glass
Specifications
SVL™ and Quickfit™ Glassware | |
Borosilicate Glass | |
Socket/screw thread ground joint adapter |