Learn More
Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stopper
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. Azlon™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
Brand:  Quickfit SB45
65.36 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12495376
  • 68.80 € / Pack of 2
Estimated Shipment: 01-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. AzlonTM Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
  • Conforms to the standard tapers as outlined in ISO 383
  • Borosilicate glass for chemical durability and high temperature performance
  • Hollow-blown construction to reduce weight
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Top of stopper is flat, allowing them to be stood on end and avoid contamination of the ground surface
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chemical durability and high temperature performance
Standard; 45/40
Stopper
Quickfit™ jointed ware and other Glassware having standard ground Glass sockets
Borosilicate Glass