Learn More
Quickfit™ Straight Tubing Adapter
Add detachable tubing to a glass vescle with a glass tubing adapter. QuickFit™ straight tubing adapters come in different sizes to fit containers or varying sizes.
Brand:  Quickfit MF13/3/SC
171.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15537930
  • 185.00 € / Pack of 5
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With straight connection for the attachment of rubber hose etc
  • Detachable plastic screwthread connectors allow flexible tubing to be fitted easily and safely
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flexible tubing, Rubber hose
Borosilicate Glass
24/29
Detachable Plastic screw thread connector
Straight tubing adapter