Learn More
Quickfit™ Unbadged Joints Cones
Free from any ceramic marking
Brand: Quickfit CNB40UB
64.03 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Free from any ceramic marking
- Avoids problem of badge charring at high temperatures which can give an untidy appearance
- Manufactured to accuracy which surpasses requirements of ISO 383
- Pyrex™ borosilicate glass for resistance to chemical corrosion and thermal shock
- Cones manufactured with shank length of 100mm
Specifications
Joint Cone | |
40/38 | |
2mm |