missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Unbadged Joints Cones

Free from any ceramic marking

Brand:  Quickfit CNB12UB

 View more versions of this product

Product Code. 15597760

  • 387.00 € / Pack of 50
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Free from any ceramic marking

  • Avoids problem of badge charring at high temperatures which can give an untidy appearance
  • Manufactured to accuracy which surpasses requirements of ISO 383
  • Pyrex™ borosilicate glass for resistance to chemical corrosion and thermal shock
  • Cones manufactured with shank length of 100mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10Pack
Male Ground Joint
12/21
1.5mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.