missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Vacuum Receiver Adapter

Deliver liquids between glass containers under vacuum. QuickFit™ vacuum receiver adapter comes with a side arm for easy attachement to vacuum supply.

Brand:  Quickfit RA3/23/SC

 View more versions of this product

Product Code. 11392095

  • 81.45 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Deliver liquids between glass containers under vacuum. QuickFit™ vacuum receiver adapter comes with a side arm for easy attachement to vacuum supply.

  • Delivery stem within cone
  • With sidearm for connection to a vacuum supply
  • Easy connection to flexible tubing via detachable plastic screwthread connector
TRUSTED_SUSTAINABILITY
Specifications

Specifications

24/29
5.51in.
Detachable Plastic screw thread connector, delivery stem, side-arm
Vacuum receiver angled adapter
Flexible tubing
140mm
Borosilicate Glass
19/26
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.