Learn More
Contec™ Quiltec™ I Mop Head
BIOPHARMA
Contec Quiltec I Mop Heads are manufactured from 100% 2-ply polyester around a rayon core so are ideal for even the highest grade of cleanroom as they generate very small numbers of particle and fibres.
183.00 € - 225.00 €
Specifications
Color Code White
Product Type Mop Head
Autoclavable Not Autoclavable
Disposable Single-use
Color-coded No
View More Specs
Products 2
Product Code Brand Sterility Price Quantity & Availability  
Product Code Brand Sterility Price Quantity & Availability  
15815597
BIOPHARMA
View Documents
Contec™
MHQT0100
Non-sterile
183.00 €
Pack of 40
In Stock
to see stock.
Add to basket
15817044
BIOPHARMA
View Documents
Contec™
MHQT0150
Sterile
225.00 €
Pack of 40
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
Description
Description
  • Two-ply 100% knitted polyester over a rayon core
  • Gamma irradiated at no less than 25 kGy
  • 5 mops double bagged
  • Approx 18-20 sqm coverage
  • Certificates of conformity supplied with every batch
Specifications
Specifications
White
Not Autoclavable
No
Mop Head
Single-use
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Contec™ Quiltec™ I Mop Head