missing translation for 'onlineSavingsMsg'
Learn More

Quinhydrone, 98%, Thermo Scientific Chemicals

Product Code. 10405861
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

Quinhydrone, 98%, ACROS Organics, Quantity: 100g, Packaging: Glass bottle, Green, Melting Point: 168 deg.C to 172 deg.C, Molecular Weight: 218.21, Percent Purity: 98%, Assay Percent Range: 48.0 to 52% Hydroquinone (HPLC), CAS: 106-34-3, ChEBI: CHEBI:26491, Density: 1.4000g/mL

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-34-3
Molecular Formula C12H10O4
Molecular Weight (g/mol) 218.21
MDL Number MFCD00010310
InChI Key BDJXVNRFAQSMAA-UHFFFAOYSA-N
Synonym quinhydrone, green hydroquinone, p-benzoquinhydrone, chinhydron, chinhydron czech, unii-p4a66lq3qj, hydroquinone, compd. with p-benzoquinone, p-benzoquinone, compd. with hydroquinone, 2,5-cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol 1:1, p-benzoquinone-hydroquinone compound 1:1
PubChem CID 7801
ChEBI CHEBI:26491
IUPAC Name benzene-1,4-diol;cyclohexa-2,5-diene-1,4-dione
SMILES OC1=CC=C(O)C=C1.O=C1C=CC(=O)C=C1

Specifications

Melting Point 168°C to 172°C
Color Green
Density 1.4000g/mL
Infrared Spectrum Authentic
Assay Percent Range 48.0 to 52% Hydroquinone (HPLC)
Packaging Glass bottle
Quantity 500 g
Merck Index 15, 8174
Specific Gravity 1.4
Solubility Information Solubility in water: 4g/L (20°C). Other solubilities: soluble in hot water,ammonia,alcohol,ether,insoluble in petroleum ether
Formula Weight 218.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Quinhydrone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Quinhydrone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.