Learn More
Quinine, 98+%, anhydrous, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 176.0°C to 177.0°C |
Color | White |
Loss on Drying | 1% max. (105°C, 2 hrs) |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Quantity | 10 g |
Fieser | 06,501; 07,311; 08,340; 09,403; 10,338; 11,456 |
Merck Index | 15, 8177 |
Specific Rotation Condition | − 172.00 (20.00°C c=1,C2H5OH) |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0424900 | Certificate of Analysis | 25/08/2023 | 163700500,163700100,163702500 |
A0401786 | Certificate of Analysis | 15/12/2022 | 163700500,163700100,163702500 |
A0396279 | Certificate of Analysis | 14/12/2022 | 163700500,163700100,163702500 |
A0412897 | Certificate of Analysis | 13/12/2022 | 163700500,163700100,163702500 |
Safety and Handling


- Quinine, anhydrous
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.