missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Quinine sulfate dihydrate, 99+% Product Code.: 10743161

Thermo Scientific Chemicals Quinine sulfate dihydrate, 99+%

Product Code. 10743161
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10743161

Brand: Thermo Scientific Chemicals 163710100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6119-70-6
Molecular Formula C40H54N4O10S
Molecular Weight (g/mol) 782.95
MDL Number MFCD00150790
InChI Key ZHNFLHYOFXQIOW-OIGVVMIYNA-N
PubChem CID 134129495
IUPAC Name (R)-[(2R,4R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;tetrahydrate
SMILES O.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=CShow MoreShow Less

Specifications

Melting Point 233°C to 235°C
Color Beige to Brown or Yellow
Ignition Residue 0.1 % max.
Loss on Drying 3% to 5% (105°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 10 g
Merck Index 15, 8177
Specific Rotation Condition −245.00 (20.00°C c=2,0.1M HCl)
Specific Rotation -245
Solubility Information Solubility in water: 1g/810mL (20°C). Other solubilities: 1g/120mL alcohol,slightly soluble in chloroform and ether
Formula Weight 782.96
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Quinine sulfate dihydrate, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459484 Certificate TypeCertificate of Analysis Date23/01/2024 Catalog Number163710100,163710500
Lot NumberA0448837 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number163710100,163710500
Lot NumberA0441874 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog Number163710100,163710500
Lot NumberA0402945 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number163710100,163710500
Lot NumberA0415014 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number163710100,163710500
compliance-icons
Product Identifier
  • Quinine sulfate dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Quinine sulfate dihydrate, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.