missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-3-boronic acid, 97%, Thermo Scientific Chemicals

84.80 € - 1139.00 €

Chemical Identifiers

CAS 191162-39-7
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.98
MDL Number MFCD02183527
InChI Key YGDICLRMNDWZAK-UHFFFAOYSA-N
Synonym 3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl
PubChem CID 2734663
IUPAC Name quinolin-3-ylboronic acid
SMILES OB(O)C1=CC2=CC=CC=C2N=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10733023
View Documents Promotion Details
Thermo Scientific Acros
367730010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10437592
View Documents Promotion Details
Thermo Scientific Acros
367730050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11945851
View Documents Promotion Details
Thermo Scientific Acros
367730250
25 g Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

191162-39-7
172.98
YGDICLRMNDWZAK-UHFFFAOYSA-N
2734663
OB(O)C1=CC2=CC=CC=C2N=C1
C9H8BNO2
MFCD02183527
3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl
quinolin-3-ylboronic acid

Specifications

191162-39-7
Brown to Orange or Pink
97%
MFCD02183527
YGDICLRMNDWZAK-UHFFFAOYSA-N
quinolin-3-ylboronic acid
2734663
97%
Quinoline-3-boronic acid
148.0°C to 155.0°C
Authentic
C9H8BNO2
3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl
OB(O)C1=CC2=CC=CC=C2N=C1
172.98
172.98
Powder
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

Product Identifier
  • 3-Quinoline boronic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only