missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-3-boronic acid hydrochloride, 97%, Thermo Scientific™

Product Code. 15429797
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-71-7
Molecular Formula C9H9BClNO2
Molecular Weight (g/mol) 209.44
MDL Number MFCD06659836
InChI Key BQFQABXCFQWPGH-UHFFFAOYSA-N
Synonym quinolin-3-ylboronic acid hydrochloride, quinoline-3-boronic acid hydrochloride, 3-quinolineboronic acid hydrochloride, quinoline-3-boronic acid hcl, quinoline-3-boronic acid, hcl, 3-quinolineboronic acid, hcl, quinoline-3-boronic acid,hydrochloride, pubchem5872, 3-quinolinylboronic acid hydrochloride
PubChem CID 44118768
IUPAC Name quinolin-3-ylboronic acid;hydrochloride
SMILES Cl.OB(O)C1=CN=C2C=CC=CC2=C1

Specifications

Melting Point 190°C to 194°C
Quantity 1 g
Formula Weight 209.44
Percent Purity 97%
Chemical Name or Material Quinoline-3-boronic acid hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.