Learn More
Quinoline-4-carboxaldehyde, 97+%, Thermo Scientific Chemicals
86.40 € - 373.86 €
Chemical Identifiers
CAS | 4363-93-3 |
---|---|
Molecular Formula | C10H7NO |
Molecular Weight (g/mol) | 157.17 |
MDL Number | MFCD00006781 |
InChI Key | MGCGJBXTNWUHQE-UHFFFAOYSA-N |
Synonym | 4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde |
PubChem CID | 78072 |
IUPAC Name | quinoline-4-carbaldehyde |
SMILES | O=CC1=C2C=CC=CC2=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309046
|
Thermo Scientific Alfa Aesar
L20203.06 |
5 g |
86.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319046
|
Thermo Scientific Alfa Aesar
L20203.14 |
25 g |
373.86 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store under dry inert gas. Incompatible with oxidizing agents and air.
Chemical Identifiers
4363-93-3 | |
157.17 | |
MGCGJBXTNWUHQE-UHFFFAOYSA-N | |
78072 | |
O=CC1=C2C=CC=CC2=NC=C1 |
C10H7NO | |
MFCD00006781 | |
4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde | |
quinoline-4-carbaldehyde |
Specifications
4363-93-3 | |
131°C to 133°C (5 mmHg) | |
C10H7NO | |
113072 | |
4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde | |
MGCGJBXTNWUHQE-UHFFFAOYSA-N | |
quinoline-4-carbaldehyde | |
78072 | |
≥97% |
48°C to 52°C | |
>110°C (230°F) | |
MFCD00006781 | |
Air Sensitive | |
Slightly soluble in water. | |
O=CC1=C2C=CC=CC2=NC=C1 | |
157.17 | |
157.17 | |
Quinoline-4-carboxaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 224-453-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only