missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinoline, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -15.0°C |
Color | Brown to Yellow |
Density | 1.0950g/mL |
Boiling Point | 237.0°C |
Flash Point | 101°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.6230 to 1.6260 |
Quantity | 500 mL |
Show More |
Safety and Handling
Product Identifier
- Quinoline
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 2
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H411-Toxic to aquatic life with long lasting effects.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362-Take off contaminated clothing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'