missing translation for 'onlineSavingsMsg'
Learn More

(-)-Quinpirole hydrochloride, Tocris Bioscience™

Product Code. 30001183
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective D2-like agonist

Selective dopamine D2 receptor agonist (Ki values are 4.8, ∽24, ∽30 and 1900 nM at D2, D3, D4 and D1 receptors respectively).

Chemical Identifiers

CAS 85798-08-9
Molecular Formula C13H22ClN3
Molecular Weight (g/mol) 255.79
InChI Key HJHVRVJTYPKTHX-HTMVYDOJSA-N
Synonym --quinpirole hydrochloride, quinpirole hydrochloride, quinpirole hcl, unii-t6i2w5v2k1, quinpirole hydrochloride usan, dsstox_cid_25519, dsstox_rid_80926, dsstox_gsid_45519, --4ar,8ar-4,4a,5,6,7,8,8a,9-octahydro-5-propyl-1h-pyrazolo 3,4-g quinoline monohydrochloride
PubChem CID 55397
IUPAC Name (4aR,8aR)-5-propyl-1,4,4a,6,7,8,8a,9-octahydropyrazolo[3,4-g]quinoline;hydrochloride
SMILES CCCN1CCCC2C1CC3=C(C2)NN=C3.Cl

Specifications

Quantity 10 mg
Formula Weight 255.79
Percent Purity >98%
Chemical Name or Material (-)-Quinpirole hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.