missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-1-Phenyl-1,3-propanediol, 98%, Thermo Scientific Chemicals
168.00 € - 526.00 €
Chemical Identifiers
CAS | 103548-16-9 |
---|---|
Molecular Formula | C9H12O2 |
Molecular Weight (g/mol) | 152.19 |
MDL Number | MFCD00145213 |
InChI Key | RRVFYOSEKOTFOG-SECBINFHSA-N |
Synonym | r-+-1-phenyl-1,3-propanediol, r-1-phenylpropane-1,3-diol, r-1-phenyl-1,3-propanediol, 1r-1-phenylpropane-1,3-diol, r-1-phenyl-1,3-dihydroxypropane, r-3-phenyl-1,3-dihydroxypropane, 1,3-propanediol,1-phenyl-, 1r, r ?-?1-?phenyl-?1,?3-?propanediol, r-+-1-phenyl-1,3-propanediol sum of enantiomers, gc |
PubChem CID | 2735120 |
IUPAC Name | (1R)-1-phenylpropane-1,3-diol |
SMILES | C1=CC=C(C=C1)C(CCO)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10712792
|
Thermo Scientific Acros
417750010 |
1 g | Glass bottle |
168.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10413575
|
Thermo Scientific Acros
417750050 |
5 g | Glass bottle |
526.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
103548-16-9 | |
152.19 | |
RRVFYOSEKOTFOG-SECBINFHSA-N | |
2735120 | |
C1=CC=C(C=C1)C(CCO)O |
C9H12O2 | |
MFCD00145213 | |
r-+-1-phenyl-1,3-propanediol, r-1-phenylpropane-1,3-diol, r-1-phenyl-1,3-propanediol, 1r-1-phenylpropane-1,3-diol, r-1-phenyl-1,3-dihydroxypropane, r-3-phenyl-1,3-dihydroxypropane, 1,3-propanediol,1-phenyl-, 1r, r ?-?1-?phenyl-?1,?3-?propanediol, r-+-1-phenyl-1,3-propanediol sum of enantiomers, gc | |
(1R)-1-phenylpropane-1,3-diol |
Specifications
103548-16-9 | |
100.0 | |
White | |
97.5% min. (GC) | |
MFCD00145213 | |
+ 54.5 | |
RRVFYOSEKOTFOG-SECBINFHSA-N | |
(1R)-1-phenylpropane-1,3-diol | |
2735120 | |
98% | |
(R)-1-Phenyl-1, 3-propanediol |
97.5 | |
62°C to 66°C | |
Authentic | |
C9H12O2 | |
+54.50° (22°C c=1,1,2−dichloroeth) | |
r-+-1-phenyl-1,3-propanediol, r-1-phenylpropane-1,3-diol, r-1-phenyl-1,3-propanediol, 1r-1-phenylpropane-1,3-diol, r-1-phenyl-1,3-dihydroxypropane, r-3-phenyl-1,3-dihydroxypropane, 1,3-propanediol,1-phenyl-, 1r, r ?-?1-?phenyl-?1,?3-?propanediol, r-+-1-phenyl-1,3-propanediol sum of enantiomers, gc | |
C1=CC=C(C=C1)C(CCO)O | |
152.19 | |
152.19 | |
Crystalline Solid |
Safety and Handling
Product Identifier
- (R)-1-Phenyl-1,3-propanediol
RUO – Research Use Only