missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenylethanol, 99%, ee 97+%, Thermo Scientific Chemicals

Product Code. 11364186
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(R)-(+)-1-Phenylethanol is used in the synthesis of optically active products. It is also useful in the determination of enantiomeric purity and for resolutions of acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-Phenylethanol is used in the synthesis of optically active products. It is also useful in the determination of enantiomeric purity and for resolutions of acids.

Solubility
Miscible with organic solvents and slightly miscible with water.

Notes
Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1517-69-7
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00064263
InChI Key WAPNOHKVXSQRPX-UHFFFAOYNA-N
Synonym r-1-phenylethanol, r-+-1-phenylethanol, 1r-1-phenylethanol, r-alpha-methylbenzyl alcohol, r-+-sec-phenethyl alcohol, 1r-1-phenylethan-1-ol, r-+-1-phenylethyl alcohol, 1-phenylethanol, r, benzenemethanol, alpha-methyl-, r
PubChem CID 637516
ChEBI CHEBI:45616
SMILES CC(O)C1=CC=CC=C1

Specifications

Melting Point 9°C to 11°C
Density 1.012
Boiling Point 88°C to 89°C (10 mmHg)
Flash Point 85°C (185°F)
Odor Mild
Refractive Index 1.528
Quantity 25 g
UN Number UN2937
Beilstein 2039798
Solubility Information Miscible with organic solvents and slightly miscible with water.
Optical Rotation +45° (c=5 in Methanol)
IUPAC Name (1R)-1-phenylethanol
Formula Weight 122.17
Percent Purity 99%
Assay ee ≥97%
Chemical Name or Material (R)-(+)-1-Phenylethanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.