missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(1-Hydroxyethyl)pyridine, Thermo Scientific Chemicals

Product Code. 15434909
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

For synthesis of optically active products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For synthesis of optically active products.

Solubility
Soluble in ethanol and water.

Notes
Store in a tightly closed container.
TRUSTED_SUSTAINABILITY

Specifications

CAS 27911-63-3
Color Colorless
Molecular Formula C7H9NO
MDL Number MFCD04972322
Quantity 1 g
Beilstein 3537186
Synonym r-1-pyridin-2-yl ethanol, r-2-1-hydroxyethyl pyridine, r-1-2-pyridyl ethanol, 1r-1-pyridin-2-yl ethan-1-ol, r-alpha-methyl-2-pyridinemethanol, 1r-1-pyridin-2-ylethanol, 1r-1-pyridin-2-yl ethanol, pubchem5699, r-1-pyridin-2-yl-ethanol, 1r-1-2-pyridyl ethanol
Solubility Information Soluble in ethanol and water.
InChI Key PPHIIIRFJKDTLG-ZCFIWIBFSA-N
SMILES CC(C1=CC=CC=N1)O
IUPAC Name (1R)-1-pyridin-2-ylethanol
Molecular Weight (g/mol) 123.155
PubChem CID 642847
Formula Weight 123.16
Physical Form Solid
Chemical Name or Material (R)-2-(1-Hydroxyethyl)pyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'