missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-1-butanol, 98%

Product Code. 10793644 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 10793644

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5856-63-3 | C4H11NO | 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5856-63-3
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00064419
InChI Key JCBPETKZIGVZRE-UHFFFAOYNA-N
Synonym r-2-aminobutan-1-ol, r---2-amino-1-butanol, 2r-2-aminobutan-1-ol, r-2-amino-1-butanol, 1-butanol, 2-amino-, 2r, unii-4f725v55ln, 2-amino-1-butanol, r, 2-amino-1-butanol,-, l---2-amino-1-butanol, 1-butanol, 2-amino-, r
PubChem CID 2723856
IUPAC Name 2-aminobutan-1-ol
SMILES CCC(N)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -2°C
Color White
Density 0.9470g/mL
Boiling Point 172°C to 174°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(NH2)CH2OH
Refractive Index 1.4495 to 1.4535
Quantity 25 mL
Beilstein 04,III,771
Merck Index 10, 429
Specific Gravity 0.947
Specific Rotation Condition −10° (19°C c=neat)
Specific Rotation -10
Solubility Information Solubility in water: miscible. Other solubilities: very soluble in most of the usual solvents,hardly soluble in aliphatic hydrocarbons
Formula Weight 89.14
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material (R)-(-)-2-Amino-1-butanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.