Learn More
(R)-(-)-2-Aminooctane, 98+%, ee 98+%, Thermo Scientific Chemicals
42.10 € - 418.00 €
Chemical Identifiers
CAS | 34566-05-7 |
---|---|
Molecular Formula | C8H19N |
Molecular Weight (g/mol) | 129.247 |
MDL Number | MFCD01091017 |
InChI Key | HBXNJMZWGSCKPW-MRVPVSSYSA-N |
Synonym | r-2-aminooctane, r-2-octylamine, 2r-2-octanamine, 2-octanamine, 2r, r---2-aminooctane, unii-rjm666k1lk, 2r-octan-2-amine, rjm666k1lk, l-2-aminooctane, --2-octylamine |
PubChem CID | 2734519 |
IUPAC Name | (2R)-octan-2-amine |
SMILES | CCCCCCC(C)N |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34566-05-7 | |
129.247 | |
HBXNJMZWGSCKPW-MRVPVSSYSA-N | |
2734519 | |
CCCCCCC(C)N |
C8H19N | |
MFCD01091017 | |
r-2-aminooctane, r-2-octylamine, 2r-2-octanamine, 2-octanamine, 2r, r---2-aminooctane, unii-rjm666k1lk, 2r-octan-2-amine, rjm666k1lk, l-2-aminooctane, --2-octylamine | |
(2R)-octan-2-amine |
Specifications
34566-05-7 | |
0.77 | |
50°C (122°F) | |
1.4235 | |
UN2734 | |
Air Sensitive | |
HBXNJMZWGSCKPW-MRVPVSSYSA-N | |
(2R)-octan-2-amine | |
2734519 | |
≥99% | |
(R)-(-)-2-Aminooctane |
<-20°C | |
163°C to 165°C | |
C8H19N | |
MFCD01091017 | |
1719320 | |
r-2-aminooctane, r-2-octylamine, 2r-2-octanamine, 2-octanamine, 2r, r---2-aminooctane, unii-rjm666k1lk, 2r-octan-2-amine, rjm666k1lk, l-2-aminooctane, --2-octylamine | |
CCCCCCC(C)N | |
129.247 | |
129.25 | |
ee ≥99% |
Safety and Handling
GHS H Statement
H300-H314-H318-H226
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H301-H314-H335
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.