Learn More
(R)-(-)-2-(Anilinomethyl)pyrrolidine, 98%, Thermo Scientific™
70.20 € - 199.00 €
Chemical Identifiers
CAS | 68295-45-4 |
---|---|
Molecular Formula | C11H16N2 |
Molecular Weight (g/mol) | 176.263 |
MDL Number | MFCD01862117 |
InChI Key | MCHWKJRTMPIHRA-LLVKDONJSA-N |
Synonym | r-n-pyrrolidin-2-ylmethyl aniline, r---2-anilinomethylpyrrolidine, r---2-anilinomethyl pyrrolidine, n-2r-pyrrolidin-2-yl methyl aniline, n-2r-pyrrolidin-2-ylmethyl aniline, r-2-anilinomethyl pyrrolidine, r-2-phenylaminomethyl-pyrrolidine, r-2-phenylamino-methyl pyrrolidine |
PubChem CID | 6950383 |
IUPAC Name | N-[[(2R)-pyrrolidin-2-yl]methyl]aniline |
SMILES | C1CC(NC1)CNC2=CC=CC=C2 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used in the determination of the absolute configuration of amines and α-amino acids by 1h nmr of (r)-o-aryllactic acid amides.
Solubility
Not miscible or difficult to mix in water.
Notes
Air sensitive. Store away from air, carbon dioxide and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
68295-45-4 | |
176.263 | |
MCHWKJRTMPIHRA-LLVKDONJSA-N | |
6950383 | |
C1CC(NC1)CNC2=CC=CC=C2 |
C11H16N2 | |
MFCD01862117 | |
r-n-pyrrolidin-2-ylmethyl aniline, r---2-anilinomethylpyrrolidine, r---2-anilinomethyl pyrrolidine, n-2r-pyrrolidin-2-yl methyl aniline, n-2r-pyrrolidin-2-ylmethyl aniline, r-2-anilinomethyl pyrrolidine, r-2-phenylaminomethyl-pyrrolidine, r-2-phenylamino-methyl pyrrolidine | |
N-[[(2R)-pyrrolidin-2-yl]methyl]aniline |
Specifications
68295-45-4 | |
120°C to 122°C (2 mmHg) | |
C11H16N2 | |
MFCD01862117 | |
4839749 | |
r-n-pyrrolidin-2-ylmethyl aniline, r---2-anilinomethylpyrrolidine, r---2-anilinomethyl pyrrolidine, n-2r-pyrrolidin-2-yl methyl aniline, n-2r-pyrrolidin-2-ylmethyl aniline, r-2-anilinomethyl pyrrolidine, r-2-phenylaminomethyl-pyrrolidine, r-2-phenylamino-methyl pyrrolidine | |
MCHWKJRTMPIHRA-LLVKDONJSA-N | |
C1CC(NC1)CNC2=CC=CC=C2 | |
176.263 | |
176.26 | |
(R)-(-)-2-(Anilinomethyl)pyrrolidine |
1.046 | |
>110°C (230°F) | |
1.575 | |
UN2735 | |
Air Sensitive | |
Not miscible or difficult to mix in water. | |
−20° (c=1 in Ethanol) | |
N-[[(2R)-pyrrolidin-2-yl]methyl]aniline | |
6950383 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
Your input is important to us. Please complete this form to provide feedback related to the content on this product.