Learn More
(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98+%, Thermo Scientific Chemicals
40.00 € - 377.00 €
Chemical Identifiers
CAS | 84025-81-0 |
---|---|
Molecular Formula | C6H13NO |
Molecular Weight (g/mol) | 115.176 |
MDL Number | MFCD00066219 |
InChI Key | CHPRFKYDQRKRRK-ZCFIWIBFSA-N |
Synonym | r-2-methoxymethyl pyrrolidine, 2r-2-methoxymethyl pyrrolidine, r---2-methoxymethyl pyrrolidine, o-methyl-d-prolinol, r-2-methoxymethyl-pyrrolidine, r-2-methoxymethylpyrrolidine, pyrrolidine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl-pyrrolidine, r-+-2-methoxymethyl pyrrolidine, h-pro-ol-me |
PubChem CID | 671216 |
IUPAC Name | (2R)-2-(methoxymethyl)pyrrolidine |
SMILES | COCC1CCCN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358846
|
Thermo Scientific Alfa Aesar
L19492.MD |
250 mg |
40.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338846
|
Thermo Scientific Alfa Aesar
L19492.03 |
1 g |
104.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348846
|
Thermo Scientific Alfa Aesar
L19492.06 |
5 g |
377.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
84025-81-0 | |
115.176 | |
CHPRFKYDQRKRRK-ZCFIWIBFSA-N | |
671216 | |
COCC1CCCN1 |
C6H13NO | |
MFCD00066219 | |
r-2-methoxymethyl pyrrolidine, 2r-2-methoxymethyl pyrrolidine, r---2-methoxymethyl pyrrolidine, o-methyl-d-prolinol, r-2-methoxymethyl-pyrrolidine, r-2-methoxymethylpyrrolidine, pyrrolidine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl-pyrrolidine, r-+-2-methoxymethyl pyrrolidine, h-pro-ol-me | |
(2R)-2-(methoxymethyl)pyrrolidine |
Specifications
84025-81-0 | |
61°C to 62°C (40 mmHg) | |
C6H13NO | |
MFCD00066219 | |
4229755 | |
r-2-methoxymethyl pyrrolidine, 2r-2-methoxymethyl pyrrolidine, r---2-methoxymethyl pyrrolidine, o-methyl-d-prolinol, r-2-methoxymethyl-pyrrolidine, r-2-methoxymethylpyrrolidine, pyrrolidine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl-pyrrolidine, r-+-2-methoxymethyl pyrrolidine, h-pro-ol-me | |
−2.2° (c=2 in Benzene) | |
(2R)-2-(methoxymethyl)pyrrolidine | |
671216 | |
≥98% |
0.933 | |
45°C (113°F) | |
1.4457 | |
UN1993 | |
Air Sensitive | |
CHPRFKYDQRKRRK-ZCFIWIBFSA-N | |
COCC1CCCN1 | |
115.176 | |
115.18 | |
(R)-(-)-2-(Methoxymethyl)pyrrolidine |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.