missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98%, Thermo Scientific Chemicals

84.10 € - 298.00 €

Chemical Identifiers

CAS 84025-81-0
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
InChI Key CHPRFKYDQRKRRK-ZCFIWIBFSA-N
Synonym r-2-methoxymethyl pyrrolidine, 2r-2-methoxymethyl pyrrolidine, r---2-methoxymethyl pyrrolidine, o-methyl-d-prolinol, r-2-methoxymethyl-pyrrolidine, r-2-methoxymethylpyrrolidine, pyrrolidine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl-pyrrolidine, r-+-2-methoxymethyl pyrrolidine, h-pro-ol-me
PubChem CID 671216
IUPAC Name (2R)-2-(methoxymethyl)pyrrolidine
SMILES COCC1CCCN1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10200800
View Documents
Thermo Scientific Acros
376190010
1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10862641
View Documents
Thermo Scientific
376190050
5 mL
298.00 €
5mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

84025-81-0
115.17
r-2-methoxymethyl pyrrolidine, 2r-2-methoxymethyl pyrrolidine, r---2-methoxymethyl pyrrolidine, o-methyl-d-prolinol, r-2-methoxymethyl-pyrrolidine, r-2-methoxymethylpyrrolidine, pyrrolidine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl-pyrrolidine, r-+-2-methoxymethyl pyrrolidine, h-pro-ol-me
(2R)-2-(methoxymethyl)pyrrolidine
C6H13NO
CHPRFKYDQRKRRK-ZCFIWIBFSA-N
671216
COCC1CCCN1

Specifications

84025-81-0
100.0
0.9320g/mL
45°C
97.5% min. (GC)
0.932
r-2-methoxymethyl pyrrolidine, 2r-2-methoxymethyl pyrrolidine, r---2-methoxymethyl pyrrolidine, o-methyl-d-prolinol, r-2-methoxymethyl-pyrrolidine, r-2-methoxymethylpyrrolidine, pyrrolidine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl-pyrrolidine, r-+-2-methoxymethyl pyrrolidine, h-pro-ol-me
CHPRFKYDQRKRRK-ZCFIWIBFSA-N
(2R)-2-(methoxymethyl)pyrrolidine
671216
98%
(R)-(-)-2-(Methoxymethyl)pyrrolidine
97.5
Colorless
62.0°C (40.0 mmHg)
Authentic
C6H13NO
-32.5° to -36.5° (20°C, 589nm) (c=2 in aq HCl)
Solubility in water: soluble
COCC1CCCN1
115.17
115.17
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapour.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

Product Identifier
  • (R)-(-)-2-(Methoxymethyl)pyrrolidine, 98%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only