missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Phenylpropionic acid, 97%, Thermo Scientific Chemicals

Product Code. 11488277
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-26-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00063140
InChI Key YPGCWEMNNLXISK-SSDOTTSWSA-N
Synonym r---2-phenylpropionic acid, r-2-phenylpropanoic acid, 2r-2-phenylpropanoic acid, r-hydratropic acid, --hydratropic acid, r---hydratropic acid, r---2-phenylpropionicacid, r-2-phenylpropionic acid, r-2-phenyl-propionic acid, r-2-phenyl-proprionic acid
PubChem CID 446626
ChEBI CHEBI:43035
IUPAC Name (2R)-2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 32°C to 34°C
Density 1.1
Boiling Point 264°C to 266°C
Flash Point 148°C (298°F)
Refractive Index 1.523
Quantity 0.25 g
Beilstein 2207688
Optical Rotation −73° (c=1.6 in Chloroform)
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material (R)-(-)-2-Phenylpropionic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'