Learn More
(R)-(-)-2-(Trifluoromethyl)pyrrolidine, 95%, Thermo Scientific™
111.00 € - 320.00 €
Chemical Identifiers
CAS | 1073556-31-6 |
---|---|
Molecular Formula | C5H8F3N |
Molecular Weight (g/mol) | 139.121 |
MDL Number | MFCD07784180 |
InChI Key | VINAMCOZNJHNIH-SCSAIBSYSA-N |
Synonym | r---2-trifluoromethyl pyrrolidine, r-2-trifluoromethyl pyrrolidine, r-2-trifluoromethylpyrrolidine, 2 r-2-trifluoromethylpyrrolidine, 2r-2-trifluoromethyl pyrrolidine, r-2-trifluoromethyl-pyrrolidine, 2-r-2-trifluoromethylpyrrolidine, 2 r-2-trifluoromethyl pyrrolidine, 2r-2-trifluoromethyl-pyrrolidine |
PubChem CID | 7015082 |
IUPAC Name | (2R)-2-(trifluoromethyl)pyrrolidine |
SMILES | C1CC(NC1)C(F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1073556-31-6 | |
139.121 | |
VINAMCOZNJHNIH-SCSAIBSYSA-N | |
7015082 | |
C1CC(NC1)C(F)(F)F |
C5H8F3N | |
MFCD07784180 | |
r---2-trifluoromethyl pyrrolidine, r-2-trifluoromethyl pyrrolidine, r-2-trifluoromethylpyrrolidine, 2 r-2-trifluoromethylpyrrolidine, 2r-2-trifluoromethyl pyrrolidine, r-2-trifluoromethyl-pyrrolidine, 2-r-2-trifluoromethylpyrrolidine, 2 r-2-trifluoromethyl pyrrolidine, 2r-2-trifluoromethyl-pyrrolidine | |
(2R)-2-(trifluoromethyl)pyrrolidine |
Specifications
1073556-31-6 | |
110°C to 112°C | |
C5H8F3N | |
UN2925 | |
r---2-trifluoromethyl pyrrolidine, r-2-trifluoromethyl pyrrolidine, r-2-trifluoromethylpyrrolidine, 2 r-2-trifluoromethylpyrrolidine, 2r-2-trifluoromethyl pyrrolidine, r-2-trifluoromethyl-pyrrolidine, 2-r-2-trifluoromethylpyrrolidine, 2 r-2-trifluoromethyl pyrrolidine, 2r-2-trifluoromethyl-pyrrolidine | |
−11° (c=1 in Methanol) | |
(2R)-2-(trifluoromethyl)pyrrolidine | |
7015082 | |
95% |
27°C to 31°C | |
26°C (79°F) | |
MFCD07784180 | |
Air Sensitive | |
VINAMCOZNJHNIH-SCSAIBSYSA-N | |
C1CC(NC1)C(F)(F)F | |
139.121 | |
139.12 | |
(R)-(-)-2-(Trifluoromethyl)pyrrolidine |
Safety and Handling
GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 623-378-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.