Learn More
Alfa Aesar™ (R)-(+)-3-Chloro-1-phenyl-1-propanol, 98%
27.70 € - 308.00 €
Chemical Identifiers
CAS | 100306-33-0 |
---|---|
Molecular Formula | C9H11ClO |
Molecular Weight (g/mol) | 170.636 |
MDL Number | MFCD00075128 |
InChI Key | JZFUHAGLMZWKTF-SECBINFHSA-N |
Synonym | r-3-chloro-1-phenylpropan-1-ol, r-+-3-chloro-1-phenyl-1-propanol, r-+-3-chloro-1-phenylpropanol, 1r-3-chloro-1-phenylpropan-1-ol, 1r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenyl-1-propanol, r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenylpropanol, pubchem5738 |
PubChem CID | 642409 |
IUPAC Name | (1R)-3-chloro-1-phenylpropan-1-ol |
SMILES | C1=CC=C(C=C1)C(CCCl)O |
Description
Chemical Identifiers
100306-33-0 | |
170.636 | |
JZFUHAGLMZWKTF-SECBINFHSA-N | |
642409 | |
C1=CC=C(C=C1)C(CCCl)O |
C9H11ClO | |
MFCD00075128 | |
r-3-chloro-1-phenylpropan-1-ol, r-+-3-chloro-1-phenyl-1-propanol, r-+-3-chloro-1-phenylpropanol, 1r-3-chloro-1-phenylpropan-1-ol, 1r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenyl-1-propanol, r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenylpropanol, pubchem5738 | |
(1R)-3-chloro-1-phenylpropan-1-ol |
Specifications
100306-33-0 | |
C9H11ClO | |
5250766 | |
JZFUHAGLMZWKTF-SECBINFHSA-N | |
C1=CC=C(C=C1)C(CCCl)O | |
170.636 | |
170.64 | |
(R)-(+)-3-Chloro-1-phenyl-1-propanol |
58°C to 60°C | |
MFCD00075128 | |
r-3-chloro-1-phenylpropan-1-ol, r-+-3-chloro-1-phenyl-1-propanol, r-+-3-chloro-1-phenylpropanol, 1r-3-chloro-1-phenylpropan-1-ol, 1r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenyl-1-propanol, r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenylpropanol, pubchem5738 | |
+26° (c=1 in Chloroform) | |
(1R)-3-chloro-1-phenylpropan-1-ol | |
642409 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.