missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-4-Benzyl-2-oxazolidinone, 99%, Thermo Scientific Chemicals

Product Code. 11472788
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(R)-(+)-4-Benzyl-2-oxazolidinones are used in the enantioselective synthesis of beta-lactams and alpha-amino acids. End products can include asysmmetric single isomeric drugs of antitumor, anesthetic, antipasmodic products. Used in the synthesis of HIV protease inhibitors. An oxazolidinone used in chiral auxiliary asymetric synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-4-Benzyl-2-oxazolidinones are used in the enantioselective synthesis of beta-lactams and alpha-amino acids. End products can include asysmmetric single isomeric drugs of antitumor, anesthetic, antipasmodic products. Used in the synthesis of HIV protease inhibitors. An oxazolidinone used in chiral auxiliary asymetric synthesis.

Solubility
Insoluble in water. Soluble in Chloroform.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102029-44-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00010846
InChI Key OJOFMLDBXPDXLQ-SECBINFHSA-N
Synonym r-4-benzyl-2-oxazolidinone, r-+-4-benzyl-2-oxazolidinone, r-4-benzyloxazolidin-2-one, 4r-4-benzyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4r, r---4-benzyl-2-oxazolidinone, r-4-benzyl-oxazolidin-2-one, 4-benzyl-1,3-oxazolidin-2-one #
PubChem CID 2734969
IUPAC Name (4R)-4-benzyl-1,3-oxazolidin-2-one
SMILES O=C1N[C@H](CC2=CC=CC=C2)CO1

Specifications

Melting Point 86°C to 89°C
Beilstein 4782551
Solubility Information Insoluble in water. Soluble in Chloroform.
Formula Weight 177.2
Percent Purity 99%
Quantity 25 g
Optical Rotation +64° (c=1 in Chloroform)
Sensitivity Hygroscopic
Chemical Name or Material (R)-(+)-4-Benzyl-2-oxazolidinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'