Learn More
Tocris Bioscience™ Blasticidin S HCl
Antibiotic; selection reagent for bis, bsr and BSD transformed cells
Brand:  Tocris Bioscience™ 5502/25
Product Code. 18785986
  • 199.00 € / 25mg
Explore more special offers
Description
Description
Blasticidin S HCl is an antibiotic. Selection reagent for bis, bsr and BSD transformed cells.
Specifications
Specifications
Blasticidin S HCl
(S)-4-[[3-Amino-5-[(aminoiminomethyl)methylamino]-1-oxopentyl]amino]-1-(4-amino-2-oxo-1(2H)-pyrimidinyl)-1,2,3,4-tetradeoxy-β-D-erythro-hex-2-enopyranuronic acid hydrochloride
Soluble to 10mM in water
458.9
Bioactive Small Molecule
3513-03-9
25 mg
C17H26N8O5·HCl
Solid
>98%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research Use Only. Not For Human or Veterinary Use