Learn More
R&D Systems™ Boc-QAR-AMC Fluorogenic Peptide Substrate
Substrate for matriptase, spinesin, and prostasin
Brand:  R&D Systems™ ES014
Product Code. 18360088
  • 336.00 € / 10mg
Explore more special offers
Description
Description
  • Hydrolysis of Arg-AMC amide bond releases AMC, a highly fluorescent group
  • A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
  • Purity >98%
  • Excellent substrate for several serine proeases
Specifications
Specifications
Boc-Gln-Ala-Arg-AMC HCl
Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for three months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.