Learn More
R&D Systems™ Boc-VPR-AMC Fluorogenic Peptide Substrate
Excellent substrate for many proteases
Brand:  R&D Systems™ ES011
Product Code. 18137250
  • 273.00 € / 20mg
Explore more special offers
Description
Description
  • Substrate for Coagulation Factor II, Coagulation Factor VII, Kallikrein 4, Kallikrein 5, Kallikrein 8, Kallikrein 13, Kallikrein 14, and MASP3
  • Hydrolysis of Arg-AMC amide bond releases AMC, a highly fluorescent group
  • A fluorescence plate reader with excitation at 380nmand emission at 460nmis recommended for the measurement of the enzymatic activity
  • Purity >95% based on TLC
Specifications
Specifications
Boc-Val-Pro-Arg-AMC
Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.