Learn More
R&D Systems™ Caspase-1/ICE Inhibitor Z-WEHD-FMK
Used as cell-permeable, irreversible inhibitor of caspase activity. R&D Systems Caspase-1/ICE Inhibitor Z-WEHD-FMK binds to the active site of activated proteases irreversibly and inhibits apoptosis.
Brand:  R&D Systems™ FMK002
Product Code. 18168590
  • 347.00 € / 1mg
Explore more special offers
Description
Description
  • Fluoromethyl ketone (FMK)-derivatized peptides are useful tools for studying caspase activity
  • Used in tissue cultures to inhibit apoptosis at final working concentrations of 50nM to 100μM
  • Regulated by a variety of stimili, including APAF1, CFLAR/FLIP, NOL3/ARC, IAP, BIRC1/NAIP, BIRC2/cIAP-1, BIRC3/cIAP-2, BIRC4/XIAP, BIRC5/Survivin and BIRC7/Livin
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Caspase-1
Z-WEHD-FMK
1 mg
877Da
Lyophilized
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For research use only. Not for use in diagnostic procedures