Learn More
R&D Systems™ Caspase Inhibitor Control Z-FA-FMK
Designed for in vitro use as a cell permeable control for the effects of fluoromethyl-ketone derivatives on caspase activity
Brand:  R&D Systems™ FMKC01
273.86 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18371698
  • 359.00 € / 5mg
Explore more special offers
Description
Description
Cell permeable fluoromethyl ketone (FMK)-derivatized peptides act as effective irreversible Caspase inhibitors with no cytotoxic effects and, therefore, are useful tools for studying Caspase activity.
  • Treated cells can then be assayed for evidence of apoptosis inhibition by examining either whole cells or cell lysates using standard apoptosis assays
  • Reconstitute in highly pure (ACS grade) DMSO fore use
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
5 mg
Caspase Inhibitor Control Z-FA-FMK, 386Da