Learn More
R&D Systems™ CellXVivo Human B Cell Expansion Kit
For the proliferative expansion of human B cell populations.
Brand:  R&D Systems™ CDK005
Product Code. 30001717
  • 502.00 € / 1 set
Explore more special offers
Includes:
  • The following optimized reagents for the expansion of human B cell populations: B Cell Expander Reagents, Reconstitution Buffers
  • Provides enough reagents for the 3-5 fold expansion of a starting population of 107 human B cells.
Specifications
Store the unopened product at 2 - 8° C. Do not use past expiration date.
1 Kit