Learn More
R&D Systems™ Cultrex RGF Basement Membrane Extract with Phenol Red
soluble form of basement membrane purified from Engelbreth-Holm-Swarm (EHS) tumor
Brand:  R&D Systems™ 3433-005-01P
228.94 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18412370
  • 291.00 € / 5mL
Explore more special offers
Description
Description
Functional Assay - Tube formation assay: BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. Gelling Assay - BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.
Specifications
Specifications
Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
≤8EU/mL
5 mL
12 to 18 mg/mL
Bioactivity
Murine Engelbreth-Holm-Swarm (EHS) tumor.