Learn More
R&D Systems™ DLDVPIPGRFDRRVS(PO3)VAAE Ser/Thr Phosphatase Substrate I
Phosphopeptide substrate for Calcineurin, PP1, PP2A, PP2C, and Ser/Thr phosphatases
Brand:  R&D Systems™ ES012
229.80 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18349687
Description
Description
  • Synthetic phosphopeptide corresponding to amino acids 81 to 99 of Protein Kinase A Regulatory Subunit 2-alpha
  • Enzymatic cleavage of this substrate liberates inorganic phosphate, which can be measured in dye binding assays, such as the malachite green method
  • Purity 95% based on HPLC analysis
Specifications
Specifications
Lyophilized
Theoretical Molecular weight: 2192 Da
No Stability Data Storage is set to N/A for ProdCode
Lyophilized from a stock solution in distilled water.
Asp-Leu-Asp-Val-Pro-Ile-Pro-Gly-Arg-Phe-Asp-Arg-Arg-Val-Ser(PO3)-Val-Ala-Ala-Glu